Durashell C18 HPLC Columns, Bonna-Agela Technologies
Supplier: Phenomenex
DC950302-0 DC950502-0 DC951502-0 DC951005-0 DC950505-0 DC951505-0 DC952505-0 DC930302-0 DC930502-0 DC930505-0 DC931002-0 DC931005-0 DC931502-0 DC931505-0 DC952510-0 DC952520-0 DC951020-0 DC951530-0 DC952530-0 DC902520-0 DC902550-0
89213-312EA 956.54 USD
89213-312 89213-314 89213-318 89213-322 89213-324 89213-326 89213-332 89213-342 89213-348 89213-350 89213-352 89213-354 89213-356 89213-358 89213-362 89213-368 89213-370 89213-374 89213-376 89213-400 89213-414
Durashell C18 HPLC Columns, Bonna-Agela Technologies
Chromatography Columns
Bonna-Agela' s Durashell column has a pH range of 1.5 to 12.0.
  • Metal Impurity: <30 ppm
  • Pore Size: 100 Å
  • Specific Surface Area: 380 m²/g
Durashell C18 HPLC Columns offer a wide pH range (1.5 to 12.0) and minimal silanol activity. They provide better efficiency than many other high pH compatible columns. They also offer a high loading capacity for basic compounds for preparative applications.
Differing from comparable products that can also be used at high pH, Durashell' s stationary phase is made with a dendrimer technology followed by molecularMore Product Information