OPTI-PAK® Trap Columns, 0.25 µl
Supplier: Optimize
10-03324-TC 10-03324-TD 10-03324-TE 10-03324-TF 10-03324-TG 10-03324-TA 10-03324-TB
89209-584PK 2014.23 USD
89209-584 89209-586 89209-588 89209-590 89209-592 89209-634 89209-636
OPTI-PAK® Trap Columns, 0.25 µl
Chromatography Columns
The OPTI-PAK®'s unique hardware design integrates a PEEK holder with an auto-adjusting stem which provides a fluid interface with any 10-32 standard injector port, and guarantees a zero-dead-volume connection despite variances in tube stop depth.
  • Specially designed for use in today's low volume trapping applications
  • Packed stem is positioned directly in flow path
  • Absolute minimum sample dilution and dispersion
  • Auto adjusting ZDV connection
  • Amenable to any trapping technique
  • Can be installed side-by-side in same valve stator without interference