SP Bel-Art Reusable Snap-on Wire Rack I.D. Tag, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F13446-0000
89209-580CS 778.93 USD
89209-580
SP Bel-Art Reusable Snap-on Wire Rack I.D. Tag, Bel-Art Products, a part of SP
Labels
Durable, reusable and easy to attach Scienceware® wire rack.
  • White coloring provides excellent contrast for most ink colors
  • One-piece write-on, wipe-off design
  • Economical and easy to use
  • Chemical-resistant and autoclavable at 121 °C (250 °F)
Smooth, polypropylene plastic surface allows labeling directly onto the tag with permanent marker, dry-erase marker, or a pressure-sensitive adhesive label.
Tags eliminate the challenges of labeling all wire racks, trays, baskets, and other wire-formed items made of 3 mm (¹/₈") or 5 mm (³/₁₆") wire.