PIG® DrainBlocker® Drain Covers with DuPont Elvaloy® Material, New Pig
Supplier: New Pig
PLR400 PLR401 PLR402 PLR403 PLR404 PLR405 PLR406 PLR412 PLR420 PLR430 PLR407 PLR408
89208-394EA 222.64 USD
89208-394 89208-396 89208-398 89208-400 89208-402 89208-404 89208-406 89208-408 89208-410 89208-412 89208-414 89208-416
PIG® DrainBlocker® Drain Covers with DuPont Elvaloy® Material, New Pig
Spill Control Materials
Use cover for quick spill response or long-term, outdoor deployment without worrying about degradation
Cover withstands prolonged exposure to diesel fuel and gasoline, even when completely submerged. Ideal for drain protection where liquid transfers, washdowns or other leak- and spill-prone jobs are common. Eliminates the need to frequently deploy and remove a temporary cover or permanently seal your drains. High-visibility yellow top layer draws attention to spills for increased safety.
Vinyl composite top layer resists stretching, ripping and tearing. DuPont Elvaloy technologyMore Product Information