Orion™ Versa Star Pro™ pH/ISE/Conductivity/Dissolved Oxygen Multiparameter Benchtop Meter, Themo Scientific
Supplier: Thermo Fisher Scientific
VSTAR90 VSTAR92 VSTAR91 VSTAR93 VSTAR94
89206-406EA 4522.18 USD
89206-406 89206-408 89206-410 89206-412 76076-764
Orion™ Versa Star Pro™ pH/ISE/Conductivity/Dissolved Oxygen Multiparameter Benchtop Meter, Themo Scientific
Electrochemistry Meters pH and Multi-Parameter Meters
Get premium performance and flexibility with the Orion™ Versa Star Pro™ Meter with pH/ISE, Conductivity and RDO/DO Modules to measure up to four channels at once.
  • Meters deliver rich functionality and features, and can be optimally configured to meet changing requirements
  • Interchangeable measurement modules allow multiple users to customize four separate channels to meet specific requirements
  • Take the guesswork out of meter calibration and setup with onscreen instructions available in a variety of local languages
  • Onscreen measurement stability indicator
More Product Information