Orion™ Versa Star Pro™ pH/ISE Benchtop Multiparameter Meter
Supplier: Thermo Fisher Scientific
VSTAR40A VSTAR40A2 VSTAR40B VSTAR40B2
89206-390EA 2710 USD
89206-390 89206-392 89206-394 89206-396
Orion™ Versa Star Pro™ pH/ISE Benchtop Multiparameter Meter
Electrochemistry Meters pH and Multi-Parameter Meters
Get premium performance and ultimate flexibility with the Orion™ Versa Star Pro™ Benchtop Multiparameter Meter with pH/ISE Module for pH, ion concentration, and temperature.
  • Onscreen measurement stability indicator lets users know exactly when measurements are ready
  • Auto-read mode holds a stable reading, continuous mode shows changing readings, Timed mode logs data at specific intervals, and Single-Shot mode holds a reading after a preset time is reached
  • Audible alarms automatically notify users when a set offset value, high/low limit value, and
More Product Information