Orion™ Star™ A216 pH/Dissolved Oxygen Benchtop Multiparameter Meter, Thermo Scientific
Supplier: Thermo Fisher Scientific
STARA2160 STARA2165
89206-344EA 2110 USD
89206-344 89206-346
Orion™ Star™ A216 pH/Dissolved Oxygen Benchtop Multiparameter Meter, Thermo Scientific
Electrochemistry Meters pH and Multi-Parameter Meters
Reliably measure pH, dissolved oxygen, and temperature with the Orion™ Star™ A216 pH/DO/RDO Benchtop Meter for advanced laboratory analysis and interfacing.
  • Combined convenience with the benefits of a pH meter and dissolved oxygen meter in one easy-to-use system
  • Easily view individual measurements on a full screen display and quickly toggle to a split screen display to view multiple measurements simultaneously
  • Perform up to a five point pH calibration using automatic buffer recognition of NIST or DIN buffer groups
  • Capture critical readings with measurement
More Product Information