PIG® Oil-Only Spill Kit in 50-Gallon Wheeled Overpack Salvage Drum, New Pig
Supplier: New Pig
KIT468
89205-846EA 1115.32 USD
89205-846
PIG® Oil-Only Spill Kit in 50-Gallon Wheeled Overpack Salvage Drum, New Pig
Spill Control Materials
Overpack drum container is UN Rated for shipping waste after oil spill cleanup
Overpack is X-rated in Packing Groups I, II and III for shipping spill cleanup waste by land, sea or rail. Lightweight, 100% polyethylene container resists chemicals and keeps contents clean and dry. Leakproof, twist-on lid is notched for easy removal or tightening with a 2x4 or pole. Large, color-coded label makes identification fast and accurate. Ledges molded into container make kit easy to move by forklift. Tamperproof seals help prevent pilfering of spill response supplies. UN1H2/X125/S.
The 6" olefinMore Product Information