1510 Disposable N95 Particulate Respirators, Moldex®
Supplier: Moldex/Metric
1511 1512 1513 1510 1517
89202-642CS 585.74 USD
89202-642 89202-644 89202-646 89202-640 89202-648
1510 Disposable N95 Particulate Respirators, Moldex®
Respirators Disposable Respirators Particulate Respirators
Heathcare particulate respirators and surgical masks feature a molded nose bridge with soft foam nose cushion to provide the wearer an easy, comfortable, and dependable fit.
  • 100% PVC-free
  • Dura-Mesh® shell prevents filter media contact by contaminated gloves or hands
  • Non-irritative elastic straps are color-coded and securely attached
  • Softspun® inner lining is soft to the touch
  • Fluid resistant with a bacterial filter efficiency of more than 99.9%
  • Available in 5 color-coded sizes
Can be used in surgery, hospitals, and healthcare facilities, nurse/hospiceMore Product Information