Tissue-Tek® Uni-Cassette® Biopsy Cassette Systems, Sakura® Finetek
Supplier: Sakura Finetek
4090 4045 4086 4087 4172 4174 4088
25608-758CS 1075.42 USD
25608-758 89199-432 89199-430 25608-754 25608-842 25608-846 25608-756
Tissue-Tek® Uni-Cassette® Biopsy Cassette Systems, Sakura® Finetek
Embedding Cassettes Tissue Processing Cassettes
The Tissue-Tek® Uni-Cassette® biopsy cassette system have a snap latch and hinge-lock cover to prevent the premature separation of lid and base during the processing of small biopsy specimens.
Biopsy cassettes are similar to Uni-Cassettes® but have 1mm pores instead of slots, eliminating the need to wrap most small specimens and greatly reducing the risk of specimen loss
Uni-Cassettes can be printed on with the Tissue-Tek® SmartWrite® printer and Tissue-Tek® AutoWrite® printer.
Uni-Cassettes can be printed on with the Tissue-Tek® SmartWrite® printer and Tissue-Tek® AutoWrite®More Product Information