Corning® Lambda™ Plus Starter Kit, Corning
Supplier: Corning
4069
89187-013CS 1312.68 USD
89187-013
Corning® Lambda™ Plus Starter Kit, Corning
Pipettes Micropipettes
The Lambda plus starter kit is designed for laboratories which are about to replace their old set of pipettors, and for customers who want to get a new set of pipettors which cover the most commonly used volumes.
The Lambda plus starter kit contains four Lambda Plus Single-Channel pipettors conveniently packaged with useful accessories (manual, calibration key, color-coded rings, tube of piston grease and identification stickers).
Ordering information: This starter kit includes four Lambda Plus Single Channel pipettors in these volume sizes 0.5 to 10 µl, 2.0 to 20 µl, 20 to 200 µl, andMore Product Information