ScrubKITS™, Corrosion Cleaning Kits, Foamtec International
Supplier: Foamtec International
HT4500-DM1 HT4500-DM2 HT4500-SSC2
89176-718CS 2845.06 USD
89176-718 89176-720 89200-068
ScrubKITS™, Corrosion Cleaning Kits, Foamtec International
Wipes Cleaning Kits and Pads
High precision remediation kits ease cleaning of corroded, rusted, or rouged 316 or 304 Stainless Steel.
Kits contain aggressive, medium, fine and super fine ScrubPADS, UltraSOLV® Sponges and MiraWIPE® wipers which enable operators to return rusted 304 and 316 stainless steel to a like-new condition using a fast process.
Starter kits include cleaning and polishing step-by-step instructions and training video for the removal of corrosion and preventing re-rusting. Refill kits provide larger quantities of ScrubPADS, UltraSOLV® Sponges and MiraWIPES® only.
Kits are compatible for useMore Product Information