Microflex® XCEED® XC-310 Nitrile Disposable Gloves
Supplier: Ansell Healthcare
XC-310-XS XC-310-M XC-310-L XC-310-S XC-310-XL
89174-538BX 67.66 USD
89174-538 89174-542 89174-544 89174-540 89174-546
Microflex® XCEED® XC-310 Nitrile Disposable Gloves
Gloves
Ergonomically designed to reduce hand fatigue while maintaining excellent tactile sensitivity.
  • Made without accelerators to protect from allergic reactions
  • ERGOFORM™ Technology; an ergonomic design that supports musculoskeletal health
  • Gloves offer advanced barrier integrity with a low 0.65 AQL
  • Fentanyl-tested for enhanced user protection
Microflex® XCEED® XC-310 gloves are manufactured without accelerators for low dermatitis potential. These gloves prevent instances of Type I latex allergies and Type IV chemical allergic reactions. To ensure extraMore Product Information
Order Now
SPECIFICATIONS
Ambidextrous/Hand-specific Ambidextrous
Environmentally Preferable
Disposable/Reusable Disposable
Latex/Latex-free Latex-Free
Powdered/Powder-free Powder-Free
Material Nitrile
Finish Textured fingers
Cuff Style Beaded
Cuff Length Standard
Cuff Thickness 2.4 mil
Finger Thickness 4.3 mil
Palm Thickness 2.8 mil
FDA Regulated +
Sterility Non-sterile
Anti-Static +
Food Contact Approved +
ASTM No. D6319