TearTech® 25 Holes Foam Rack, Therapak®
Supplier: Therapak, LLC
68825
89171-120CS 211.42 USD
89171-120
TearTech® 25 Holes Foam Rack, Therapak®
Mailers and Shippers
When combined with shipping boxes, specimen bags or insulated shippers, this foam tube rack provides cushioning and protection for 13x75mm tubes.
The unique die-cut construction allows for tearing away variable-sized grids to customize transport load. Overall foam block size is 150x150x100mm