ERGOMAT® Hygiene Rubber Mats, ERGOMAT
Supplier: Ergomat
EH0204-GRN EH0205-GRN EH0207-GRN EH0208-GRN EH0209-GRN EH0210-GRN EH0211-GRN EH0212-GRN EH0213-GRN EH0214-GRN EH0215-GRN EH0216-GRN EH0217-GRN EH0218-GRN EH0219-GRN EH0220-GRN EH0303-GRN EH0305-GRN EH0306-GRN EH0307-GRN EH0308-GRN EH0309-GRN EH0310-GRN EH0311-GRN EH0312-GRN EH0313-GRN EH0314-GRN EH0315-GRN EH0316-GRN EH0317-GRN EH0318-GRN EH0319-GRN EH0320-GRN EH0404-GRN EH0405-GRN EH0406-GRN EH0407-GRN EH0408-GRN EH0409-GRN EH0410-GRN EH0411-GRN EH0412-GRN EH0413-GRN EH0414-GRN EH0415-GRN EH0416-GRN EH0417-GRN EH0418-GRN EH0419-GRN EH0420-GRN EH0206-GRN EH0304-GRN EH0203-GRN
89169-508EA 390.49 USD
89169-508 89169-510 89169-512 89169-514 89169-516 89169-518 89169-520 89169-522 89169-524 89169-526 89169-528 89169-530 89169-532 89169-534 89169-536 89169-538 89169-540 89169-542 89169-544 89169-546 89169-548 89169-550 89169-552 89169-554 89169-556 89169-558 89169-560 89169-562 89169-564 89169-566 89169-568 89169-570 89169-572 89169-574 89169-576 89169-578 89169-580 89169-582 89169-584 89169-586 89169-588 89169-590 89169-592 89169-594 89169-596 89169-598 89169-600 89169-602 89169-604 89169-606 22231-140 22231-142 22231-138
ERGOMAT® Hygiene Rubber Mats, ERGOMAT
Floor Mats Traction and Anti-Fatigue Floor Mats
The ERGOMAT® Hygiene is a cone profile ergonomic mat that is well suited for use in critical environments requiring daily caustic sanitation.
  • Hygiene mats have a broad range of pH resistance (2 to 13), and tolerate a variety of chemical exposures (except oils)
  • Use in hospitals and clinics, laboratories, food processing facilities, and commercial kitchens
  • Available in 4 colors (blue, gray, red, and green) to help eliminate the potential for cross contamination issues
  • They can also be safely autoclaved (2’ x 3’ size only)
Mats are 11 mm (7/16") thickMore Product Information