SlimLine™ Mop Adapters, Micronova
Supplier: Micronova
QPSL-09 QPSL-14 QPSL-18 QDSL-09 QDSL-14 QDSL-18 QPSL-16 QPSL-PLUG QPSL-30
89140-692EA 118.66 USD
89140-692 89140-694 89140-696 11310-596 11310-598 11310-600 10153-678 76214-294 76323-718
SlimLine™ Mop Adapters, Micronova
Mops
A lightweight plastic; flat-head mop adapter for use with the SlimLine™, PadMop™, SnapMop™, FlatMop, and PocketMop series.
  • Lightweight plastic
  • Flathead mop adapter
  • Autoclavable
  • Compatible with strong disinfectants and common detergents
All frames fit in the SlimLine™ and SlimT™ bucket systems and attach to STA series of plastic handles.