ScrubWRIGHT® Pen Tool, Chamber Cleaning, Foamtec International
Supplier: Foamtec International
FTPEN-1
89133-226CS 323.31 USD
89133-226
ScrubWRIGHT® Pen Tool, Chamber Cleaning, Foamtec International
Wipes Cleaning Kits and Pads
A unique tool for cleaning small and hard to reach areas.
  • Helps reduce particles
  • Safely removes deposition from sensitive areas
  • Reduces damage to critical parts
  • Wide selection of Diamond ScrubBELTS®
  • Tool features a 6 mm tip at 15° angle
Interchangeable belt-shaped Diamond ScrubBELTS® in various grits create a versatile instrument perfect for polishing stains, scratches, and process-induced residue from scratch-sensitive surfaces. Spring adjustable for easy mounting and rotation of ScrubWRIGHT® ScrubBELT®.
Order Now
SPECIFICATIONS
Environmentally Preferable