TWIRL'EM ECOLO Biodegradable Sampling Bags, Labplas
Supplier: VWR International
89131-782 89131-784 89131-786 89131-788 89131-790 89131-794 89131-802 89131-806 89131-818 89131-826 89131-812 89131-814 89131-800 89131-810 89131-792 89131-804 89131-808 89131-820 89131-824 89131-830 89131-832 89131-834
89131-782CS 535.56 USD
89131-782 89131-784 89131-786 89131-788 89131-790 89131-794 89131-802 89131-806 89131-818 89131-826 89131-812 89131-814 89131-800 89131-810 89131-792 89131-804 89131-808 89131-820 89131-824 89131-830 89131-832 89131-834
TWIRL'EM ECOLO Biodegradable Sampling Bags, Labplas
Bags Sample Bags
These biodegradable sample bags are ideal for transportation and storage of solids, semisolids, and liquids for environmental and carcass sampling, biomedical and pharmaceutical research, quality assurance procedures, food industry applications, and clinical and veterinary medicine.
  • Made of pliant, highly resistant virgin polyethylene sterile tubing with no side seals
  • Feature a wire closure for an airtight seal
  • Wide-sealed bottoms for added leakproof protection
They are easy to use, tear off perforated top, pull tabs to open bag, fill, grab ends ofMore Product Information