Statfree® High Density Conductive Foam, Desco
Supplier: Desco
12550
89131-086EA 154.67 USD
89131-086
Statfree® High Density Conductive Foam, Desco
Mailers and Shippers
High Density Conductive Foam is antistatic and low tribocharging, which minimizes charge generation.
  • Antistatic, low tribo charging
  • Dissipative and corrosion resistant
  • Uniform protection
  • Volume Resistance: 10E3 – 10E5 ohms
  • Surface Resistance: 10E3 – 10E5 ohms
  • Made in the United States of America
Antistatic, low tribocharging minimizes charge generation; material will conduct charges if grounded. Corrosion resistant design is ideal for long-term storage. Uniform protection will not affect the solderability of device leads.
Certifications: Lead-free, RoHS compliant.