KleenGuard™ 30-06 Rx Readers Safety Glasses, Kimberly-Clark Professional
Supplier: Ansell Healthcare
19892 19894 19879 19876
67200-258EA 29.72 USD
67200-258 67200-268 67200-270 89130-944 89130-946 89130-950 89130-942
KleenGuard™ 30-06 Rx Readers Safety Glasses, Kimberly-Clark Professional
Safety Glasses
KleenGuard™ 30-06 readers safety glasses are equipped to suit those who need vision assistance and eye protection. Available in a variety of lens and frame colors and prescription strengths, they are designed for long-term comfort.
  • Clear lenses are ideal for indoor use
The 30-06 readers have stylish nylon frames, complete with self-adjusting soft rubber nose pads for comfort. The lenses are hardcoated for longer life and scratch resistance. Durable polycarbonate lenses provide 99.9% UVA/ UVB/UVC protection to help prevent eye damage like cataracts,More Product Information