VWR® Autoclavable Biohazard Bags and Containers
Supplier: VWR International
11215-680 14220-030 14220-054 14221-156 14221-164 14221-994 14221-996 89129-470 89129-472 89129-474 89130-796 11215-904 14220-026 14220-028 14220-032 14220-034 14220-046 14220-048 14220-050 14220-052 14220-066 14220-068 14220-070 89129-468
11215-680CS 588.67 USD
11215-680 14220-030 14220-054 14221-156 14221-164 14221-994 14221-996 89129-470 89129-472 89129-474 89130-796 11215-904 14220-026 14220-028 14220-032 14220-034 14220-046 14220-048 14220-050 14220-052 14220-066 14220-068 14220-070 89129-468
VWR® Autoclavable Biohazard Bags and Containers
Bags Waste Disposal Bags
Double-thick bags are ideal for sterilizing animal cages and other objects with sharp corners.
  • High-integrity polypropylene
  • Available with temperature indicator
  • Resists punctures, tears, and leaks
Ideal for processing biohazardous materials that require autoclaving prior to disposal.
Twist-tie closure allows bag to expand and vent steam during cooling. Bags resist punctures, tears, and leaks and meet the 165 g dart drop standard performed as required by ASTM D1709-98. Each bag with indicator is imprinted with the universal biohazard symbol.