VWR® Signature™ Shoe Covers made with DuPont™ Tyvek® IsoClean® Material
Supplier: VWR International
89128-790 89128-792 89128-794
89128-790CS 267.09 USD
89128-790 89128-792 89128-794
VWR® Signature™ Shoe Covers made with DuPont™ Tyvek® IsoClean® Material
Disposable Garments
Shoe covers are manufactured using a unique flash spinning process that creates an excellent barrier against nonhazardous liquids and microorganisms as well as dry particles down to 0.5µm.
  • Excellent barrier against nonhazardous liquids and microorganisms, dry particles down to 0.5µm
  • For cleanroom use, chemically and biologically inert, tear resistant, and low linting
  • Single use, serged seams and elastic tops for a secure fit
  • Gripper™ soles provide enhanced skid-resistance and durability
Designed for cleanroom use, garments are chemically andMore Product Information