VWR® TraceClean® Mayo Tall Glass Jars, Clear
Supplier: VWR International
89126-572 89126-574 89126-586 89126-588 89126-590
89126-572CS 175.46 USD
89126-572 89126-574 89126-586 89126-588 89126-590
VWR® TraceClean® Mayo Tall Glass Jars, Clear
Jars General Use Jars
These TraceClean® glass jars are used primarily for the collection of soil, sediments, and slurry media for testing Semi-volatiles, Pesticides, PCBs, Diesel Range Organics, TPH, Metals, and Phenolics.
  • Clear glass fabrication
  • Used as a containment vessel
  • Suited toward the collection of inorganic samples
Secondary uses include containment, bench vessels, and archiving
Quality Assured® with VWR® Logo Sample ID Label products, a lot certificate of analysis is generated with the analytical results obtained including the concentration reporting limitsMore Product Information