VWR® TraceClean® Straight Side Glass Jars, Clear
Supplier: VWR International
89126-568 89126-570 89126-578 89126-580 89126-582 89126-584 89126-592 89126-596 89094-235
89126-568CS 194.34 USD
89126-568 89126-570 89126-578 89126-580 89126-582 89126-584 89126-592 89126-596 89094-235
VWR® TraceClean® Straight Side Glass Jars, Clear
Jars General Use Jars
These TraceClean® glass jars are used primarily for the collection of soil, sediments, and slurry media for testing Semi-volatiles, Pesticides, PCBs, Diesel Range Organics, TPH, Metals, and Phenolics.
  • Clear glass fabrication
  • Straight-sided design
  • Used as a containment vessel
  • Suited to the collection of inorganic samples
Secondary uses include containment, bench vessels, and archiving
The 950 mL clear jar 89094-235 contains 5 mL (1:1) of the chemical preservative H2SO4.
Quality Assured® with VWR® Logo Sample ID Label products, a lot certificateMore Product Information