CELLview™ Cell Culture Dishes, Glass Bottom, Sterile, Greiner bio-one
Supplier: Greiner Bio-One
627860 627861 627870 627871 627965 627975
89125-438CS 239 USD
89125-438 89125-440 89125-442 89125-444 89135-974 89135-976
CELLview™ Cell Culture Dishes, Glass Bottom, Sterile, Greiner bio-one
Culture Dishes Petri Dishes
The CELLview™ cell culture dish is made from high-grade polystyrene combinded with an integrated glass bottom.
  • Sterilized by irradiation
  • Free of RNase, DNase, and human DNA
  • Noncytotoxic
  • Nonpyrogenic
The innovative design guarantees a single-plane, flat bottom with a consistent working distance and maximal planarity
CELLview™ combines the conveniance of a standard size 35mm disposable plastic cell culture dish with the optical quality of glass, providing superior high-resolution microscopic images of in-vitro cultivated cultures.
Glass bottomMore Product Information