Modern Round Plastic Bottles, Narrow Mouth, Natural
Supplier: C&G Containers
LPB004900000 LPB004900001 LPB008200110 LPB008900000 LPB008900001 LPB016900000 LPB032900000 LPB0162001 LPB0332000 LPB0332003 LPB0082001 LPB016100002 LPB004100011 LPB008100012 LPB016100012 LPB0332000
71000-912CS 452.22 USD
71000-912 71000-914 71000-918 71000-920 71000-922 71000-928 71000-938 89125-414 89125-418 89125-420 89125-413 30621-174 10496-892 10496-894 10496-896 89125-419
Modern Round Plastic Bottles, Narrow Mouth, Natural
Bottles Plastic Bottles
These plastic bottles are used for testing metals and a variety of inorganic parameters including alkalinity, anions, acidity, cyanide, sulfate, and hardness.
Pre-cleaned and Certified products contain the Certificate of Analysis and Cleaning Protocol Form in the product case.
A lot certificate of analysis is generated with the analytical results obtained, including the concentration reporting limits and units of measurement, for Pre-cleaned and Certified products. This certificate of analysis is included in each product case.
Wash protocols for these bottles include; A1 (NitricMore Product Information