Coupling Clamp, Stainless Steel, Circulator Hose, Ace Glass Incorporated
Supplier: Ace Glass
12187-28 12187-35
89097-268EA 369.47 USD
89097-268 89097-270
Coupling Clamp, Stainless Steel, Circulator Hose, Ace Glass Incorporated
Clamps Standard Taper and Spherical Joint Clamps
This stainless steel coupling clamp is designed for use with ACE jacketed glass pilot plant reactors and circulator hoses for popular circulating chillers
This clamp connects a [SJ] glass joint to a stainless steel [SJ] adapter of the appropriate size.
Order Now
SPECIFICATIONS
Environmentally Preferable