VWR® TraceClean® Vials, Clear
Supplier: VWR International
89093-840 89096-790 89126-532 89126-534 89126-538 89126-540 89126-542 89126-658 89126-660
89093-840CS 548.73 USD
89093-840 89096-790 89126-532 89126-534 89126-538 89126-540 89126-542 89126-658 89126-660
VWR® TraceClean® Vials, Clear
Vials
These TraceClean® glass jars are used for the collection of and testing of aqueous samples for Volatile Organic Analysis and Total Petroleum Hydrocarbons (TPH).
  • Clear glass fabrication
  • Suited for testing aqueous samples
  • Used as a containment vessel
Collection and testing of aqueous samples for Total Organic Carbon (TOC)
Quality Assured® with VWR® logo sample ID label products, a lot certificate of analysis is generated with the analytical results obtained including the concentration reporting limits and units of measurement. This certificate ofMore Product Information