VWR® Oven Thermometers
Supplier: VWR International
60803-0511 60803-0711 60803-0911
89095-812CS 288.2 USD
89095-812 89095-814 89095-816 89095-818
VWR® Oven Thermometers
Thermometers Analog Thermometers
Oven thermometers are individually serialized, and partial immersion versions feature a round glass swell to hold the thermometer in place.
  • Individually serialized
  • Lead-free white glass
  • Red organic liquid
Individually packaged in recyclable packaging with non-roll fitting.