VWR® General Purpose Liquid-In-Glass Thermometers
Supplier: VWR International
60700-0411 60700-0511 60700-0711 60700-0911 60700-1411 60700-1911 60700-0111 60700-1111 60700-1211
89095-598CS 526.07 USD
89095-598 89095-600 89095-602 89095-604 89095-606 89095-608 89095-610 89095-612 89095-614 89095-616 89095-618 89095-622 89095-626 89095-628
VWR® General Purpose Liquid-In-Glass Thermometers
Thermometers Analog Thermometers
Mercury free, these thermometers feature blue organic liquid fill against lead-free white back glass.
  • Ideal for use in chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical/medical industry, dairies, breweries and distilleries
  • Include multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability
  • Tested and inspected by our USA quality team
Should an individual Thermometer Calibration Report be required, factoryMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable