VWR® TraceClean® Vials with Chemical Preservative
Supplier: VWR International
89094-160 89094-180 89094-182 89094-172 89094-178 89094-158
89094-160CS 169.99 USD
89094-160 89094-180 89094-182 89094-172 89094-178 89094-158
VWR® TraceClean® Vials with Chemical Preservative
Vials
Type I borosilicate glass vials contain a chemical preservative to maintain sample integrity until volatile organic analysis can be performed.
  • Amber glass fabrication
  • Recommended for UV light-sensitive samples
  • Imbued with a chemical preservative
All vials are prepared in a solvent-free facility to prevent possible volatile contamination and are supplied with polypropylene open-top caps with ultrasonically bonded 3.1 mm fluoropolymer resin/silicone septa.
Vials include color-coded barcode with individual container number, preservative, and lotMore Product Information