VWR® TraceClean® Wide Mouth Jars, HDPE
Supplier: VWR International
89094-094 89094-096 89094-098 89094-100 89094-102 89094-104 89094-106 89094-108 89094-110
89094-094CS 224.25 USD
89094-094 89094-096 89094-098 89094-100 89094-102 89094-104 89094-106 89094-108 89094-110
VWR® TraceClean® Wide Mouth Jars, HDPE
Jars General Use Jars
Straight-sided jars are for collecting clays, slurries, and sediment matrices for use in geotechnical, environmental applications. Toxicity Characteristic Leaching Procedure (TCLP) metals, inorganic compounds, and benthic organism studies.
  • HDPE construction
  • Wide-mouthed design
  • Supplied with polyfoam-lined caps
Processed (PC) and Quality-Assured® (QA) jars follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers'. QA jars include a Certificate of Analysis in every case.
More Product Information