VWR® TraceClean® Wide Mouth Packers, Amber Glass
Supplier: VWR International
89093-966 89094-034 89094-038 89094-040 89094-044 89094-046 89094-048 89094-050 89094-052 89094-056 89093-962 89093-964 89094-036 89094-042 89094-054
89093-966CS 144.24 USD
89093-966 89094-034 89094-038 89094-040 89094-044 89094-046 89094-048 89094-050 89094-052 89094-056 89093-962 89093-964 89094-036 89094-042 89094-054
VWR® TraceClean® Wide Mouth Packers, Amber Glass
Bottles Water Sampling Bottles
Type III (soda-lime) glass packers are available with solid-top or open-top caps.
  • Soda-lime glass fabrication
  • Wide-mouthed design
  • Available with solid-top or open-top caps
  • Individually numbered for traceability
Solid-top caps with fluoropolymer resin liner are for use in semi-volatiles, pesticides, PCBs, and phenolics analyses.
Processed (PC) and Quality-Assured® (QA) packers follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers'. QA packers include a CertificateMore Product Information