PYREX® Distilling Flask, Vertical Type, Standard Taper 24/40 Joints, 3 Necks, Corning
Supplier: Corning
4950-5L 4950-1L 4950-2L
89091-476CS 1584.76 USD
89091-476 89090-696 89090-864
PYREX® Distilling Flask, Vertical Type, Standard Taper 24/40 Joints, 3 Necks, Corning
Flasks Distillation Flasks
PYREX® borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Central neck and two parallel side necks. Matt white labelling area. Three vertical neck flasks have two side necks with 24/40 outer standard taper joints and a center neck with a 24/40 standard taper joint. All the necks are at the same height.