PYREX® Distilling Flasks, Vertical Type, 3 Necks, [ST] Joints, Corning
Supplier: Corning
4960-500 4960-12L 4960-1L 4960-250 4960-2L
89091-432CS 1125.97 USD
89091-432 29280-442 89090-666 89090-808 89090-866 89090-932
PYREX® Distilling Flasks, Vertical Type, 3 Necks, [ST] Joints, Corning
Flasks Distillation Flasks
PYREX® borosilicate glass 3.3.
  • Excellent chemical resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
  • Made from heavy wall blanks to withstand severe service
Central neck and two parallel side necks. Matt white labelling area. Particularly suitable for use with a stirring device, condenser, or other apparatus as a unit.