PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning
Supplier: Corning
4060-6L 4060-12L 4060-50 4060-250 4060-1L 4060-2L 4060-125
29111-208CS 214.67 USD
29111-208 29111-230 89091-414 29111-045 29111-070 29111-128 29111-161 89090-642
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning
Flasks Flat Bottom Flasks
Manufactured from borosilicate glass.
  • High thermal shock resistance
  • Provide contamination free results
  • High chemical durability
Tooled mouth ensures uniform stopper fit. Florence, long neck. Normal working temperature is 230 °C (446 °F).
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental Certification I-RECs, ISO 14001 - Monterrey