Corning® Digital Hot Plates, Corning
Supplier: Corning
6797-600D 6795-600D 6796-600D 6798-600D 6797-400D 6796-400D 6798-400D 6795-400D
89090-186CS 937.8 USD
89090-186 89090-188 89090-190 89090-192 89090-204 89090-208 89090-210 89090-206
Corning® Digital Hot Plates, Corning
Heaters Hot Plates
Corning® hotplates with Pyroceram® glass-ceramic top, rugged housing and heating elements and digital temperature display.
  • Microprocessor maintains consistent and repeatable temperature settings from 5 °C (if ambient temperature is 0 °C or lower) up to 550 °C
  • Digital LED temperature display is adjustable in 5 °C increments and blinks until set temperature is reached
  • Optional external temperature controller (89090-224) truly controls temperature inside the vessel
  • Bright LED 'hot' icon lights up when top plate is over 60 °C, even when heat control is turned
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental Certification I-RECs, ISO 14001 - Reynosa