SP Bel-Art Vacuum Aspirator Bottle, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F19917-0001 F19917-0002
80089-736EA 386.03 USD
80089-736 89088-038
SP Bel-Art Vacuum Aspirator Bottle, Bel-Art Products, a part of SP
Bottles Aspirators
For easy and continuous vacuum aspiration of a variety of liquids including tissue culture media, wash solutions, supernatant fluids after centrifugation, and chemical waste.
  • Heavy-walled design
  • Polypropylene construction
  • Housed in an epoxy-coated steel wire cage
  • Flexible collection tubing
Consists of a heavy-walled polypropylene collection bottle housed in an epoxy-coated steel wire cage, a secondary vacuum trap, and flexible collection tubing.
Bottle and cap are autoclavable.
The collection bottle safely retains fluid while the attached trapMore Product Information