VWR® Sterile Sample Bags with Specimen Sponge
Supplier: VWR International
89003-806 89085-576
89003-806CS 1258.68 USD
89003-806 89085-576
VWR® Sterile Sample Bags with Specimen Sponge
Bags Sample Bags
Sterile sample bags are ideal for surface sampling in a wide range of applications.
  • Sterile
  • Includes cellulose sponge
  • Polyethylene sterile tubing
  • Write-on strip
  • Wire closure for an airtight seal
  • Wide-sealed bottoms for leakproof protection
They provide a reliable method for monitoring potentially hazardous environments for the presence of microorganisms as well as foodborne pathogens.
Each bag includes a cellulose sponge that measures 3.8Wx7.6Lx1.6Hcm (11/2x3x5/8") when hydrated. Choose from a biocide-free, dry sponge or a foil-sealed spongeMore Product Information