Vacuum Traps, O-Ring Flange Connection, Ace Glass
Supplier: Ace Glass
8755-05 8755-10 8755-13 8755-17
89084-822EA 362.68 USD
89084-822 89084-824 89084-826 89084-828
Vacuum Traps, O-Ring Flange Connection, Ace Glass
Vacuum Traps
This vacuum trap assembly has a Viton O-ring flange connection, offering flexibility and ease of assembly and disassembly for cleaning
This trap utilizes a stainless steel, locking pinch clamp. Fabricated from borosilicate glass.
Order Now
SPECIFICATIONS
Environmentally Preferable