Schlenk Pear Shaped Storage Flask with Sidearm and Stopcock, Ace Glass Incorporated
Supplier: Ace Glass
7754-11 7754-211 7754-22 7754-222 7754-05 7754-205 7754-13 7754-213 7754-45 7754-245 7754-24 7754-224 7754-07 7754-207 7754-47 7754-247 7754-26 7754-226 7754-09 7754-209
89083-634EA 193.77 USD
89083-634 89083-636 89083-638 89083-640 89083-642 89083-644 89083-646 89083-648 89083-650 89083-652 89083-654 89083-656 89083-658 89083-660 89083-662 89083-664 89083-666 89083-668 89083-670 89083-672
Schlenk Pear Shaped Storage Flask with Sidearm and Stopcock, Ace Glass Incorporated
Flasks Nitrogen Flasks
Used for storage, but may also be used for simple reactions
With standard taper 14/20 or standard taper 24/40 inner joint or #15 o-ring joint and 2 or 4 mm bore glass stopcock or 2 mm inlet valve. Fabricated from borosilicate glass.
Order Now
SPECIFICATIONS
Environmentally Preferable