Clamp, Extension, Vinylized Jaws, Ace Glass
Supplier: Ace Glass
11072-17
89083-256EA 80.97 USD
89083-256
Clamp, Extension, Vinylized Jaws, Ace Glass
Clamps General Purpose Clamps
This single-adjust clamp is designed so that the jaws open at a gradual angle, to their extreme position, without binding on the screws
The screw, wing-nut and pivot pin are nickel plated; the spring itself is stainless steel. The extension arm is a seamless tube, nickel-plated.
Ordering information: Comes with an extra set of fiberglass prong covers for temperatures above 100°C.