Clamp, Two-Prong, Ace Glass
Supplier: Ace Glass
11075-21
89083-208EA 71.16 USD
89083-208
Clamp, Two-Prong, Ace Glass
Clamps General Purpose Clamps
These medium size clamps are have two-pronged jaws designed to holding burets up to 38mm in diameter
The position of the jaws and holder are fixed. This insures that any object held in the jaws will remain permanently in a vertical position. Clamps to rods up to 19mm.