Power Hold Clamp, Ace Glass Incorporated
Supplier: Ace Glass
11082-07
89075-632EA 147.13 USD
89075-632
Power Hold Clamp, Ace Glass Incorporated
Clamps General Purpose Clamps
This clamp fits a support stand with 3/8" to 5/8" diameter shaft, and stirrers with mounting rod from 3/8" to 5/8" diameter
Stop collar included.