Clips for Instatherm® Oil Baths, Ace Glass Incorporated
Supplier: Ace Glass
9601-29 9601-30 9601-32 9601-34 9601-36
89075-334EA 73.4 USD
89075-334 89075-336 89075-338 89075-342 89075-346
Clips for Instatherm® Oil Baths, Ace Glass Incorporated
Baths Oil Baths
These clips, designed to hold thermocouples or thermometers, work with various Ace Instatherm® oil baths
The small size clips should be used with 89075-322 oil baths. The large clip should be used with 89075-384 oil baths. The extra-large clips should be used with 89075-332 oil baths. Medium clips can be used with any other size Instatherm® oil baths.
Order Now
SPECIFICATIONS
Environmentally Preferable