PIG® Burpless® One-Hand-Sealable Steel Drum Funnel, New Pig
Supplier: New Pig
DRM1125-YW-NPT
89073-811EA 605.33 USD
89073-811
PIG® Burpless® One-Hand-Sealable Steel Drum Funnel, New Pig
Funnels Filtration Funnels Drum Funnels
This funnel with latching handle easily opens and closes lid in a simple, single-handed operation.
  • Lockable to prevent unauthorized access
  • Suitable for 5 to 55 gal. steel drums and pails with 2" NPT
  • Durable, chemical-resistant powder coat helps ensure long service life
Unique burpless design vents during pouring for fast, smooth flow with no dangerous splashback. Lets you pour without venting through 3/4" bung, a possible closed container reg violation if left open. Nitrile gasket and locking mechanism, completely sealed lid to help reduce vaporMore Product Information