PIG® Heavy-Duty Maintenance Wipers, New Pig
Supplier: New Pig
WIP304 WIP325 WIP303 WIP306 WIP328 WIP329
56620-095CS 232.31 USD
56620-095 89069-046 89069-042 89069-044 89073-972 89073-974
PIG® Heavy-Duty Maintenance Wipers, New Pig
Wipes Disposable Wipes and Towels Wipes for General Applications
Strong and solvent-resistant for wiping machines and rough parts, but soft enough for cleaning skin or preparing surfaces.
  • Excellent for heavy-duty maintenance jobs like machine wipedowns and degreasing parts
  • Low-lint construction won't shed, pill, or leave behind unwanted fibers when wiping parts
  • Binder-free construction withstands solvents, degreasers, and cleaning agents
Wipers are 1/4 folded and poly wrapped for easy handling and loading. Convenient pop-up box lets you take just one wiper at a time while keeping remaining stock clean and dry. Pop-More Product Information