Pressure Bearing with Ace-Threds, 10 mm, Ace Glass Incorporated
Supplier: Ace Glass
8044-07 8044-13 8044-24 8044-55
89061-790EA 87.48 USD
89061-790 89061-792 89061-794 89061-796
Pressure Bearing with Ace-Threds, 10 mm, Ace Glass Incorporated
Stirrers Stirring Shafts
Glass bearings feature a #15 Ace-Thred at each end and glass tubing between
Designed for use with 10mm O.D. stirring shafts. A PTFE coupling, with internal O-Ring seal, connects the bottom of the bearing to either a #15 or a #25 Ace-Thred on vessels such as a pressure reactor head or one-piece pressure reactors. The top bushing also has an internal O-Ring, for an additional seal on the shaft to allow pressure reactions. Maximum operating speed is 600rpm. Coupling and bushing are each supplied with O-Rings. Glass components are fabricated from borosilicate glass.
Ordering information: More Product Information