Powder Funnels, Vertical, Compact, Screw Feed, Ace Glass Incorporated
Supplier: Ace Glass
7233-20 7233-30
89058-602EA 556.5 USD
89058-602 89058-604
Powder Funnels, Vertical, Compact, Screw Feed, Ace Glass Incorporated
Funnels Powder Funnels
A vertical, compact, screw feed funnel for the addition of powders and solids (up to 25 mesh) into reactions without seizing or binding
Features a flexible PTFE screw thread wrapped on a precision PTFE-ceramic shaft and contained within a precision bore housing. This offers a unique flexibility in that the thread can move aside temporarily; as one spacing increases, the adjacent spacing decreases, thereby maintaining an average feed rate. Completely inert materials allow flushing with solvents without fear of contamination. #15 Ace-Thred at top offers easy disassembly for cleaning. Large 1More Product Information