Flask, Freeze Drying, Lyophilization, Ace Glass
Supplier: Ace Glass
7035-03 7035-05 7035-09 7035-104 7035-11 7035-110 7035-116 7035-122 7035-128 7035-15 7035-150 7035-152 7035-160 7035-162 7035-17 7035-170 7035-21 7035-23 7035-27 7035-29 7035-34 7035-36 7035-40 7035-42 7035-46 7035-48 7035-52 7035-54 7035-56 7035-58
89057-484EA 252.52 USD
89057-484 89057-486 89057-488 89057-490 89057-492 89057-494 89057-496 89057-498 89057-500 89057-502 89057-504 89057-506 89057-508 89057-510 89057-512 89057-514 89057-516 89057-518 89057-520 89057-522 89057-524 89057-526 89057-528 89057-530 89057-532 89057-534 89057-536 89057-538 89057-540 89057-542
Flask, Freeze Drying, Lyophilization, Ace Glass
Flasks Flat Bottom Flasks
Complete "fast-freeze" flask with snap-on rubber cap and 1/2" or 3/4" O.D.
  • Fabricated from borosilicate glass
  • Flasks are designed for ease of handling and faster loading
Snap-on black rubber cap is grease free; one size fits all capacity flasks. Plain straight adapter for connecting flask cap to lyophilizer manifold is removable and fabricated from PTFE. PTFE adapter with shut-off valve also available. The size value accompanied with adapter size and style.
Plain or valve adapter, compatible with all major brand freeze drying manifold valves.More Product Information